missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plastic Carboy with Gasket Cap

Catalog No. S27494
Change view
Click to view available options
Capacity (Metric):
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S27494 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27494 Supplier Eisco™ Supplier No. EISCH0065B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made of lab quality polypropylene plastic, designed for holding large amounts of liquid regularly used in the laboratory

  • Premium polypropylene plastic carboys
  • Ideal for any laboratory
  • Gasket Lids seal the container
  • Significantly lighter than glass, highly portable, and durable
  • Handles provide easy maneuverability and placement
  • Autoclavable

Specifications

Autoclavable Autoclavable
Capacity (Metric) 20 L
Closure Material Plastic
Closure Type Screw Cap with Gasket
Color White
Disposable Reusable
For Use With (Application) Holding Liquid
Graduations No
Material Polypropylene
Mouth Style Narrow
Neck Style Short
Product Type Carboy
Shape Cylindrical
Sterility Non-sterile
Show More Show Less