missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Innovative Med Tech Plastic Blood Gas Capillary Tubes: Closing Caps
Closing caps for Innovative Med Tech Plastic Blood Gas Capillary Tubes
Supplier: Innovative Med Tech 110180
Description
- Made from 100% plastic
- Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Compliance
Meets FDA, OSHA, and CDC mandates
Chemical Identifiers
| Closing Cap | |
| Plastic Blood Gas Capillaries | |
| FDA; OSHA; CDC |
| Plastic | |
| Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening |
Specifications
| Closing Cap | |
| Plastic Blood Gas Capillaries | |
| 1.75 mm |
| Plastic | |
| Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening | |
| FDA; OSHA; CDC |