missing translation for 'onlineSavingsMsg'
Learn More

Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives

Catalog No. 22757148
Encompass
Change view
Click to view available options
Capacity (Metric):
110 μL
115 μL
130 μL
155 μL
Diameter (Metric) Inner:
78 mm
100 mm
120 mm
Length (Metric):
1.75 mm
1.9 mm
2.35 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Diameter (Metric) Inner Length (Metric)
22757148 115 μL 78 mm 1.9 mm
22757136 130 μL 120 mm 1.75 mm
22757143 155 μL 100 mm 2.35 mm
22757135 110 μL 100 mm 1.75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22757148 Supplier Innovative Med Tech Supplier No. 102117*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes

  • Made from 100% plastic
  • Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening

Compliance

Meets FDA, OSHA, and CDC mandates

Specifications

Product Type Capillary Tube
Additive Balanced Li-Heparin
Material Plastic
Capacity (Metric) 115 μL
Diameter (Metric) Inner 78 mm
For Use With (Application) Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Length (Metric) 1.9 mm
Certifications/Compliance FDA; OSHA; CDC