missing translation for 'onlineSavingsMsg'
Learn More

Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives

Catalog No. 22757136
Change view
Click to view available options
Capacity (Metric):
110 μL
115 μL
130 μL
Diameter (Metric) Inner:
78 mm
100 mm
120 mm
Length (Metric):
1.75 mm
1.9 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Diameter (Metric) Inner Length (Metric)
22757136 130 μL 120 mm 1.75 mm
22757148 115 μL 78 mm 1.9 mm
22757135 110 μL 100 mm 1.75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22757136 Supplier Innovative Med Tech Supplier No. 101120

Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes

  • Made from 100% plastic
  • Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening

Compliance

Meets FDA, OSHA, and CDC mandates

Specifications

Product Type Capillary Tube
Additive Balanced Li-Heparin
Material Plastic
Capacity (Metric) 130 μL
Diameter (Metric) Inner 120 mm
For Use With (Application) Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Length (Metric) 1.75 mm
Certifications/Compliance FDA; OSHA; CDC