Learn More
United Scientific™ Plastic Beakers
Plastic Beakers feature excellent clarity and very good chemical resistance.
Supplier:  United Scientific™ BP1000PK5
 View more versions of this product
Catalog No. S01335
Only null left
Description
Description
  • Molded in polypropylene
  • Autoclavable
  • Feature raised graduations on the outer surface
  • Griffin Style Beakers nest without sticking together
  • New Style beakers feature tapered spouts for easier pouring of solutions
Specifications
Chemical Identifiers
Polypropylene
Autoclavable
Griffin
Beaker
Yes
Specifications
33.8 oz.
Polypropylene
5 Pk.
Autoclavable
Yes
Griffin
1000 mL
Beaker for liquid measurement
Beaker
Reusable
Molded Scale
Product Suggestions
Product Suggestions