Learn More
Dynalon™ Plastic Beaker with Handle
Our handles make your handling of solutions easier and safer.
Supplier:  Dynalon™ 3264952000/E
 View more versions of this product
Catalog No. S30726
Only null left
Description
Description
  • Durable beakers feature a low form design for better stability
  • Made of autoclavable polypropylene
Specifications
Chemical Identifiers
Translucent
Handle
Autoclavable
Polypropylene
Beaker
Yes
Specifications
Translucent
14.4 cm
Polypropylene
6.5 in.
Handle
Autoclavable
5.7 in.
2000 mL
50 to 2000 mL
16.51 cm
Beaker
Yes
Product Suggestions
Product Suggestions