missing translation for 'onlineSavingsMsg'
Learn More

Youngstown Barrel & Drum Plastic 3.5 Gal TH w/70mm screw cap

Catalog No. 18120572
Change view
Click to view available options
Capacity (English):
3.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18120572 3.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18120572 Supplier Youngstown Barrel & Drum Supplier No. 2383

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3.5 Gal TH pail w/70mm screw cap. Round. HDPE. 2.4 lbs 11 in. OD x 12 in.OH. A tight head pail is often used when shipping a free liquid that requires a more secure seal at the closure.UN rated 1H1/Y1.8/100.

Seamless construction prevents leaks. Used for liquids.

Specifications

Certifications/Compliance UN 1H1/Y1.8/100
Product Type Pail
Closure Type 70 mm Screw Cap
Capacity (English) 3.5 gal.
Capacity (Metric) 15.4 L
Color White
For Use With (Application) PACKAGING FOR HAZARDOUS LIQUIDS
Material Polyethylene
Freight Class 250
Head Style Tight
Includes 70 mm Screw Cap
Length (English) 12 in.
Width (English) 11 in.
Height (English) 12 in.
Height (Metric) 30.48 cm
Diameter (English) Outer 11 in.
Diameter (Metric) Outer 27.94 mm
Weight (English) 2.4 lb.
Weight (Metric) 1.08862169 Kg
Show More Show Less