missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plas Labs™ Model 830 Compact Glove Box
For safe containment of hazardous materials when being weighed
$20593.81 - $25601.13
Specifications
| Product Type | Glove Box |
|---|---|
| Certifications/Compliance | UL, CSA, CE |
| Voltage | 120 V |
Includes: White ambidextrous Hypalon™ gloves for superior chemical and U.V. resistance; Manual.
| Catalog Number | Mfr. No. | Depth (English) | Dimensions (L x W x H) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Depth (English) | Dimensions (L x W x H) | Price | Quantity | ||||
|
11-389-6
|
Plas Labs™
830ABCEXP |
24 in. | 23 x 43 x 31 in. (61 x 110 x 79 cm) |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
11-389-121
|
Plas Labs™
830ABB |
30 in. | 30 x 49 x 31 in. (77 x 125 x 79 cm) |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
11-386-137
|
Plas Labs™
830ABE |
32 in. | 81.2 x 193 x 89 cm |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Work chamber accommodates analytical and toploading balances, bench scales and moisture analyzers in a clean, safe, portable and draft-free atmosphere.
- Top section is removable for easy installation of equipment
- Four purging gas valves (two on the transfer chamber and two on the main chamber)
- White leveling tray for easy transferring of liquids
- All clamps stainless steel and completely adjustable
- Pressure relief valve minimizes glove fight back
- HEPA filter on relief valve prevents escape of particulates and organic vapors
- HEPA and organic vapor canister filters (replaceable)
- Roomy transfer chamber is 12L x 11 in. I.D.
Certifications
Included hospital-grade multiple electrical outlet strip is UL listed, CSA approved.
Specifications
| Glove Box | |
| UL, CSA, CE | |
| 120 V |