Learn More
Plas Labs™ Flat-Bottom Rodent Restrainers
Apertures in the top and bottom provide easy access for injections or sampling
Supplier:  Plas Labs™ 543 RR
 View more versions of this product
Catalog No. 0128831C
Only null left
Description
Description
  • Pocket and holes in the front make it easy to anesthetize or feed the animal
  • Clear material provides a visual check of rodent's condition
  • Adjustable tail stock can be inserted into any one of four positions
Specifications
Chemical Identifiers
Clear
Flat Bottom Rodent Restrainer
Chemically sterilized or washed at 180°F
Injections or Blood Sampling
Noryl™ Plastic
Specifications
Clear
Flat Bottom Rodent Restrainer
Chemically sterilized or washed at 180°F
15.24 cm
6.35 cm
Injections or Blood Sampling
Noryl™ Plastic
6 in.
2.5 in.
125 to 250 g
Product Suggestions
Product Suggestions