Learn More
Plas Labs™ 856-HYPO Glove Box
Ideal for tissue culture work
Supplier:  Plas Labs™ 856HYPO
Catalog No. 11-389-132
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The Hypoxia Chamber Glove Box is ideal for tissue culture work, including tumor cell and stem cell research. It features a color operated “touch screen” for automatically controlling oxygen levels, temperature levels, and carbon dioxide levels.
  • Thermoelectric assembly (peltier device)
  • Platinum RTD with sealing gland
  • O2 sensor and CO2 sensor
  • Air circulation fan
  • Inert gas hook-up
  • N2, CO2 and O2 gas hook-up
  • Configurable for up to six individually controlled sampling chambers
  • 8 in. color panel touch screen
  • Password protected, configurable PID control
  • One touch, on-screen datalogging
  • Ethernet port connection for data logging
Specifications
Specifications
Enclosure
UL
28 in.
71 cm
38 in.
97 cm
104 cm
55 in.
139.7 cm
6.5/5 A
35 in.
89 cm
55 x 55 x 38 in. (104 x 139.7 x 97 cm)
26 in.
55 in.
115/230 V
41 in.
104.14 cm
Product Suggestions
Product Suggestions