Learn More
Brands PL AQUAGEL-OH GUARD 15UM -1/UN
Catalog No. 500106980
Encompass
Agilent LS PL aquagel-OH Guard 15um 50 x 7.5 mm LC Columns - GPC/SEC BC
Catalog No. 500106980 Supplier Brands Supplier No. PL11491240
Agilent LS PL aquagel-OH Guard 15um 50 x 7.5 mm LC Columns - GPC/SEC BC