missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pizotifen malate

Catalog No. AAH3660303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3660303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3660303 Supplier Thermo Scientific™ Supplier No. H3660303

Specifications

CAS 5189-11-7
Molecular Formula C23H27NO5S
MDL Number MFCD01700978
Quantity 1 g
InChI Key IWAWCPZVTXCFKD-UHFFFAOYNA-N
SMILES OC(CC(O)=O)C(O)=O.CN1CCC(CC1)=C1C2=C(CCC3=CC=CC=C13)SC=C2
IUPAC Name 1-methyl-4-{6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-2-ylidene}piperidine; 2-hydroxybutanedioic acid
Molecular Weight (g/mol) 429.53
Formula Weight 295.44
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 225-970-4
RTECSNumber TM7165500