missing translation for 'onlineSavingsMsg'
Learn More

Pixantrone dimaleate

Catalog No. AC465522500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465522500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC465522500 Supplier Thermo Scientific Chemicals Supplier No. 465522500
Only null left

CAS: 144675-97-8 | C25H27N5O10 | 557.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144675-97-8
Molecular Formula C25H27N5O10
Molecular Weight (g/mol) 557.52
InChI Key SVAGFBGXEWPNJC-LVEZLNDCSA-N
IUPAC Name bis((2E)-but-2-enedioic acid); 6,9-bis[(2-aminoethyl)amino]-5H,10H-benzo[g]isoquinoline-5,10-dione
SMILES OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.NCCNC1=CC=C(NCCN)C2=C1C(=O)C1=CC=NC=C1C2=O

Specifications

Alpha Vector PIXANTRONEDIMALEATE
Color Black to Purple
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 250 mg
Solubility Information (2 mg/ml aq. soln.) Clear solution
Formula Weight 557.51
Physical Form Powder
Chemical Name or Material Pixantrone dimaleate
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only