missing translation for 'onlineSavingsMsg'
Learn More

Pivaloylacetonitrile 98.0+%, TCI America™

$92.41 - $750.82

Chemical Identifiers

CAS 59997-51-2
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00010208
InChI Key MXZMACXOMZKYHJ-UHFFFAOYSA-N
Synonym pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
PubChem CID 108871
IUPAC Name 4,4-dimethyl-3-oxopentanenitrile
SMILES CC(C)(C)C(=O)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P111225G
View Documents
TCI America
P111225G
25 g
Each for $92.41
Only null left
 
P1112500G
View Documents
TCI America
P1112500G
500 g
Each for $750.82
Only null left
 
Description

Description

Specifications

Chemical Identifiers

59997-51-2
125.171
MXZMACXOMZKYHJ-UHFFFAOYSA-N
108871
CC(C)(C)C(=O)CC#N
C7H11NO
MFCD00010208
pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
4,4-dimethyl-3-oxopentanenitrile

Specifications

59997-51-2
White-Yellow
C7H11NO
25 g
pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
CC(C)(C)C(=O)CC#N
125.171
125.17
Crystalline Powder
68°C
126°C
MFCD00010208
3439
MXZMACXOMZKYHJ-UHFFFAOYSA-N
4,4-dimethyl-3-oxopentanenitrile
108871
≥98.0% (GC)
Pivaloylacetonitrile
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3720

RTECSNumber : SA2991000

TSCA : Yes

SDS
Documents

Documents