missing translation for 'onlineSavingsMsg'
Learn More

Pivaloylacetonitrile 98.0+%, TCI America™

Catalog No. P111225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111225G 25 g
P1112500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P111225G Supplier TCI America Supplier No. P111225G

Chemical Identifiers

CAS 59997-51-2
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00010208
InChI Key MXZMACXOMZKYHJ-UHFFFAOYSA-N
Synonym pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
PubChem CID 108871
IUPAC Name 4,4-dimethyl-3-oxopentanenitrile
SMILES CC(C)(C)C(=O)CC#N

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 3439
Formula Weight 125.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pivaloylacetonitrile
EINECSNumber (2)-3720
RTECSNumber SA2991000
TSCA Yes