missing translation for 'onlineSavingsMsg'
Learn More

Pivaloylacetonitrile, 97%

Catalog No. AC418025000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418025000 500 g Glass bottle
AC418020250 25 g Glass bottle
AC418021000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC418025000 Supplier Thermo Scientific Chemicals Supplier No. 418025000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59997-51-2 | C7H11NO | 125.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59997-51-2
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.17
MDL Number MFCD00010208
InChI Key MXZMACXOMZKYHJ-UHFFFAOYSA-N
Synonym pivaloylacetonitrile, trimethylacetylacetonitrile, 4,4-dimethyl-3-oxovaleronitrile, pentanenitrile, 4,4-dimethyl-3-oxo, 4,4-dimethyl-3-oxo-pentanenitrile, cyanopinacolone, pivaloyl acetonitrile, 1-cyano-3,3-dimethyl-2-butanone, valeronitrile, 4,4-dimethyl-3-oxo, pyvaolylacetonitrile
PubChem CID 108871
IUPAC Name 4,4-dimethyl-3-oxopentanenitrile
SMILES CC(C)(C)C(=O)CC#N

Specifications

Melting Point 66°C to 72°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC(O)CH2CN
Quantity 500 g
Formula Weight 125.17
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pivaloylacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 262-017-1

RUO – Research Use Only