missing translation for 'onlineSavingsMsg'
Learn More

Pivalonitrile 98.0+%, TCI America™

Catalog No. P046325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P046325ML 25 mL
P0463100ML 100 mL
P0463500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P046325ML Supplier TCI America Supplier No. P046325ML

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 mL
UN Number 1993
Formula Weight 83.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pivalonitrile