missing translation for 'onlineSavingsMsg'
Learn More

Pivalic Anhydride 98.0+%, TCI America™

Catalog No. p-7135125
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P141425ML 25 mL
P1414250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P141425ML Supplier TCI America Supplier No. P141425ML
Only null left

Chemical Identifiers

CAS 1538-75-6
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00008842
InChI Key PGZVFRAEAAXREB-UHFFFAOYSA-N
Synonym pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
PubChem CID 15234
IUPAC Name 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC(=O)C(C)(C)C

Specifications

Color Colorless
Quantity 25 mL
UN Number 2920
Formula Weight 186.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pivalic Anhydride