missing translation for 'onlineSavingsMsg'
Learn More

Pivalic Acid 99.0+%, TCI America™

Catalog No. p-7134620
Change view
Click to view available options
Quantity:
25 g
100 g
400 g
Catalog No. Quantity
P046125G 25 g
P0461100G 100 g
P0461400G 400 g
3 options

Catalog No. P046125G

Supplier: TCI America P046125G

Only null left

Chemical Identifiers

CAS 75-98-9
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004194
InChI Key IUGYQRQAERSCNH-UHFFFAOYSA-N
Synonym pivalic acid, trimethylacetic acid, neopentanoic acid, 2,2-dimethylpropionic acid, tert-pentanoic acid, propanoic acid, 2,2-dimethyl, versatic 5, kyselina pivalova, acetic acid, trimethyl, alpha,alpha-dimethylpropionic acid
PubChem CID 6417
ChEBI CHEBI:45133
IUPAC Name 2,2-dimethylpropanoic acid
SMILES CC(C)(C)C(=O)O

Specifications

Color White
Boiling Point 164°C
Quantity 25 g
UN Number 3261
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Pivalic Acid
EINECSNumber (2)-0608
RTECSNumber TO7700000
TSCA Yes