missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pivalic Acid 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 164°C |
| Quantity | 100 g |
| UN Number | 3261 |
| Formula Weight | 102.13 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Pivalic Acid |
Safety and Handling
| EINECSNumber | (2)-0608 |
| RTECSNumber | TO7700000 |
| TSCA | Yes |