missing translation for 'onlineSavingsMsg'
Learn More

Pivalamidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135241
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17615G 5 g
P176125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17615G Supplier TCI America Supplier No. P17615G

Chemical Identifiers

CAS 18202-73-8
Molecular Formula C5H13ClN2
Molecular Weight (g/mol) 136.623
MDL Number MFCD00051988
InChI Key ARDGQYVTLGUJII-UHFFFAOYSA-N
Synonym 2,2-dimethylpropanimidamide hydrochloride, pivalamidine hydrochloride, pivalimidamide hydrochloride, tert-butylcarbamidine hydrochloride, 2,2-dimethylpropionamidine hydrochloride, 2,2-dimethyl-propionamidine hydrochloride, 2,2-dimethylpropionamidine hcl, t-butylcarbamidine hydrochloride, 2,2-dimethylpropanamidine hydrochloride, 2,2,2-trimethylacetamidine hydrochloride
PubChem CID 2781880
IUPAC Name 2,2-dimethylpropanimidamide;hydrochloride
SMILES CC(C)(C)C(=N)N.Cl

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
Formula Weight 136.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Pivalamidine Hydrochloride