missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Pivalamide | |
| 154°C | |
| 212°C | |
| MFCD00008011 | |
| pivalamide, trimethylacetamide, 2,2,2-trimethylacetamide, propanamide, 2,2-dimethyl, 2,2-dimethylpropionamide, 2,2-dimethyl-propionamide, unii-fes86mr7zi, fes86mr7zi, propanamide,2,2-dimethyl, trimethylacetarnide | |
| CC(C)(C)C(N)=O | |
| 101.15 | |
| 101.15 | |
| Crystalline Powder |
| 754-10-9 | |
| White-Yellow | |
| C5H11NO | |
| 100 g | |
| XIPFMBOWZXULIA-UHFFFAOYSA-N | |
| 2,2-dimethylpropanamide | |
| 12957 | |
| ≥98.0% (GC,N) |
Chemical Identifiers
| 754-10-9 | |
| 101.15 | |
| XIPFMBOWZXULIA-UHFFFAOYSA-N | |
| 12957 | |
| CC(C)(C)C(N)=O |
| C5H11NO | |
| MFCD00008011 | |
| pivalamide, trimethylacetamide, 2,2,2-trimethylacetamide, propanamide, 2,2-dimethyl, 2,2-dimethylpropionamide, 2,2-dimethyl-propionamide, unii-fes86mr7zi, fes86mr7zi, propanamide,2,2-dimethyl, trimethylacetarnide | |
| 2,2-dimethylpropanamide |
Safety and Handling
TSCA : Yes