missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine dihydrochloride, 99%

Catalog No. AAJ62252MC
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62252MC 100 mg
AAJ6225203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62252MC Supplier Thermo Scientific Chemicals Supplier No. J62252MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A M1 muscarinic receptor antagonist | CAS: 29868-97-1 | C19H23Cl2N5O2 | 424.326 g/mol

A M1 muscarinic receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29868-97-1
Molecular Formula C19H23Cl2N5O2
Molecular Weight (g/mol) 424.326
MDL Number MFCD00055214
InChI Key FFNMBRCFFADNAO-UHFFFAOYSA-N
Synonym pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
PubChem CID 71405
ChEBI CHEBI:32014
IUPAC Name 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
SMILES CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl

Specifications

Melting Point 192°C to 198°C
Color White
Quantity 100 mg
Sensitivity Hygroscopic
Merck Index 14,7491
Formula Weight 424.32
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Pirenzepine dihydrochloride
EINECSNumber 249-907-5
RTECSNumber UU7883000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only