missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PIPPS, Calbiochem™,

Catalog No. 528315100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
528315100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 528315100GM Supplier MilliporeSigma™ Supplier No. 528315100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A non-metal complexing, zwitterionic buffer with mixed-mode dissociation constants of pKa1 = 3.73 and pKa2 = 7.96 (100mM, 25°C)

PIPPS has no interference at wavelengths longer than 240nm

Specifications

CAS 5625-56-9
Quantity 100 g
Synonym Piperazine-N,N'-bis(3-propanesulfonic Acid)
Molecular Weight (g/mol) 330.414