Learn More
VistaLab™ Technologies Pipetter Nozzles
Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter
$126.03 - $340.21
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|
22-146-664
|
VistaLab™ Technologies
90573017 |
Pack of 10 Each for $158.47
|
|
||||||
|
22-146-660
|
VistaLab™ Technologies
90572010 |
100 to 200 μL |
Pack of 10 Each for $273.12
|
|
|||||
|
22-146-653
|
VistaLab™ Technologies
90571010 |
2 to 20 μL |
Pack of 10 Each for $273.12
|
|
|||||
|
22-146-656
|
VistaLab™ Technologies
90571013 |
2 μL |
Pack of 5 Each for $340.21
|
|
|||||
|
22-146-306
|
VistaLab™ Technologies
90605010 |
5 mL |
Pack of 2 Each for $126.03
|
|
|||||
Description
- Available in range of sizes
Specifications
| Pipettes | |
| Plastic |