missing translation for 'onlineSavingsMsg'
Learn More

Poltex Pipette Tip Receptacle

Catalog No. 22000760
Encompass
Change view
Click to view available options
Includes:
Magnet Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22000760 Wall Mount
22000758 Magnet Mount
22000759 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22000760 Supplier Poltex Supplier No. TIPDISPW
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Receptacle for used pipette tips.

  • Back plate mounts to wall, inside of hood, etc.
  • Front receptacle lifts off for easy emptying/cleaning.
  • Choose from VHB tape mount (very high bond tape included), or magnet mount (powerful magnets included).

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipettes
Material PETG
Positions 1
Shape Rectangular, Rounded Corners
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipette
Length (Metric) 11 cm
Includes Wall Mount
Product Type Pipet Rack
Array 1 x 1
Show More Show Less