Learn More
United Scientific™ Pipette Stand, Horizontal, 12-Place, Plastic
Catalog No. S15854
Catalog No. S15854 Supplier United Scientific™ Supplier No. 79101
Only null left
Safely store your glass pipettes.
  • This polypropylene pipette stand can hold up to 12 pipettes in a horizontal position.
  • Constructed with three polypropylene coated metal rods for extra weight and stability
  • Easily disassembled and reassembled
  • Autoclavable
Specifications
Autoclavable Autoclavable
Color White
Holds Pipettes
Material Polypropylene
Shape Vertical
Stackable No
Sterility Non-sterile
Includes 6 Racks
Product Type Pipette Stand
Shelves None