missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAXTM MAX1000 Pipette Head

Catalog No. FC10010
Change view
Click to view available options
Volume (Metric):
100 to 1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
FC10010 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FC10010 Supplier Gilson™ Supplier No. FC10010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pipetting head with a volume range of 100 to 1000 μm.

The PIPETMAXTM offers 8-channel pipette heads with the system. Choose from three pipetting volume options:

  • 1 to 10 μL
  • 20 to 200 μL
  • Fixed 1 mL

Specifications

Product Type Pipetter Accessory
Certifications/Compliance ISO 9001
Channels 1
Material Polypropylene
Volume (Metric) 100 to 1000 μL
For Use With (Application) Liquid Handling
For Use With (Equipment) PIPETMAX™
Includes 1 Cassette B1000
Software No
Tip Material Polypropylene
Width (English) 10.5 in
Voltage 0 to 100 mV, 1 V, 10 V
Show More Show Less