Learn More
Gilson™ PIPETMAXTM 8-channel Pipette Head
Catalog No. FC10022
Change view
Click to view available options
Volume (Metric):
1 to 20 μL
20 to 200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
FC10022 1 to 20 μL
FC10021 20 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FC10022 Supplier Gilson™ Supplier No. FC10022
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pipetting head with 8 channels for use with its automated pipettor.
The PIPETMAXTM offers 8-channel pipette heads with the system. Choose from different pipetting volume options:
  • 1 to 10 μL
  • 20 to 200 μL
Specifications
Certifications/Compliance ISO 9001
Channels 8
Material Polypropylene
Volume (Metric) 1 to 20 μL
For Use With (Application) Liquid Handling
For Use With (Equipment) PIPETMAX
Includes 1 Cassette B8x20
Software No
Tip Material Polypropylene
Width (English) 10.5 in
Voltage 0 to 100 mV, 1 V, 10 V
Show More Show Less