Learn More
Pipette Supplies Inc PIPETMAN TIP EJECTOR P1000
Supplier: Pipette Supplies Inc GIF123659
Tip ejector for:Gilson Pipetman Classic 1000L (P1000)Gilson Pipetman Fixed F250, F300, F400, F500, F1000Gilson Pipetman Neo 1000L (P1000N)Made in France
Description
Greener Choice Claims
- Environmental benefits include:
- Source Reduction