missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PipetmanTM EXPERTTM Empty Racks for Standard Pipette Tips
GREENER_CHOICE

Catalog No. F1735021G
Change view
Click to view available options
Holds:
Pipette Tips (10μL)
Pipette Tips (1000μL)
Pipette Tips (200μL)
Pipette Tips (300μL)
Quantity:
1 Rack/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Quantity
F1735021G Pipette Tips (1000μL) 1 Rack/Pk.
F1732021G Pipette Tips (10μL) 1 Rack/Pk.
F1734021G Pipette Tips (300μL) 1 Rack/Pk.
F1733021G Pipette Tips (200μL) 1 Rack/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. F1735021G Supplier Gilson™ Supplier No. F1735021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For holding up to 1000μL pipette tips

For use with EXPERT Universal fit tips.

Specifications

Clean Claims RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free
Autoclavable Autoclavable
Color Clear
Holds Pipette Tips (1000μL)
Material Polypropylene
Positions 1
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) EXPERT Universal Fit Pipette Tips
Quantity 1 Rack/Pk.
Size 229 x 159 x 229 mm
Length (English) 5 in.
Width (English) 3.5 in.
Height (English) 4.35 in.
Includes Empty Rack
Product Type Pipette Tip Rack
Array 1x1
Show More Show Less