missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PipetmanTM EXPERTTM Empty Racks for Standard Pipette Tips
GREENER_CHOICE

Catalog No. F1734021G
Change view
Click to view available options
Holds:
Pipette Tips (10μL)
Pipette Tips (1000μL)
Pipette Tips (200μL)
Pipette Tips (300μL)
Quantity:
1 Rack/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Quantity
F1734021G Pipette Tips (300μL) 1 Rack/Pk.
F1732021G Pipette Tips (10μL) 1 Rack/Pk.
F1735021G Pipette Tips (1000μL) 1 Rack/Pk.
F1733021G Pipette Tips (200μL) 1 Rack/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. F1734021G Supplier Gilson™ Supplier No. F1734021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For holding up to 1000μL pipette tips

For use with EXPERT Universal fit tips.

Specifications

Autoclavable Autoclavable
Color Clear
Holds Pipette Tips (300μL)
Material Polypropylene
Positions 1
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) EXPERT Universal Fit Pipette Tips
Quantity 1 Rack/Pk.
Size 140 x 108 x 70 mm
Length (English) 5 in.
Width (English) 3.5 in.
Height (English) 2.75 in.
Includes Empty Rack
Product Type Pipette Tip Rack
Array 1x1
Show More Show Less