missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM DIAMONDTM Tips Reload Pack
GREENER_CHOICE

Catalog No. F167014G
Change view
Click to view available options
Model:
D1000
D200
DL10
Volume (Metric):
0.1 to 20 μL
100 to 1000 μL
2 to 200 μL
Color Code:
Dark Blue
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Volume (Metric) Color Code
F167014G D1000 100 to 1000 μL Dark Blue
F167012G DL10 0.1 to 20 μL Red
F167013G D200 2 to 200 μL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F167014G Supplier Gilson™ Supplier No. F167014
Only null left

PIPETMANTM DIAMONDTM Tips Reload Pack is an easy and quick way to refill an empty TIPACKTM Box. Three times smaller than a regular tips box, it is easy to store on your bench.

Accurate and Precise

  • Using PIPETMANTM DIAMONDTM Tips with your PIPETMANTM ensures maximum accuracy and precision and delivers consistent results.
  • The perfect air-tight seal prevents leaks.
  • The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing.
  • Volume graduations make it easy to visually check the volume aspirated.

Reliable

  • GilsonTM tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your results.
  • From mold to bench, our tips are guaranteed, certified and traceable to meet the most stringent demands in many applications.

Convenient Packaging

  • Using up to 70% less plastic than a regular tip box, GilsonTM's reload pack is an eco-friendly and affordable solution.
  • Its small size makes it easy to store.
  • The racks are fully autoclavable and are color-coded for easy identification and compliance with Good Laboratory Practice (GLP).

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model D1000
Length (English) 2.60 in.
Length (Metric) 66 mm
Certifications/Compliance ISO 9001
For Use With (Application) Refill empty TIPACK™ boxes
Diameter (Metric) Outer .85 mm
Format Refill
Tip Style Wide Bore
Packaging Reload pack
Quantity 96 Tips/Pk.
Product Type Universal Pipette Tips
Color Code Dark Blue
Disposable Single-use
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman Diamond
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less