missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM DIAMONDTM Tips Reload Pack
GREENER_CHOICE

Catalog No. F167012G
Change view
Click to view available options
Model:
D1000
D200
DL10
Volume (Metric):
0.1 to 20 μL
100 to 1000 μL
2 to 200 μL
Color Code:
Dark Blue
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Volume (Metric) Color Code
F167012G DL10 0.1 to 20 μL Red
F167013G D200 2 to 200 μL Yellow
F167014G D1000 100 to 1000 μL Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F167012G Supplier Gilson™ Supplier No. F167012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PIPETMANTM DIAMONDTM Tips Reload Pack is an easy and quick way to refill an empty TIPACKTM Box. Three times smaller than a regular tips box, it is easy to store on your bench.

Accurate and Precise

  • Using PIPETMANTM DIAMONDTM Tips with your PIPETMANTM ensures maximum accuracy and precision and delivers consistent results.
  • The perfect air-tight seal prevents leaks.
  • The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing.
  • Volume graduations make it easy to visually check the volume aspirated.

Reliable

  • GilsonTM tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your results.
  • From mold to bench, our tips are guaranteed, certified and traceable to meet the most stringent demands in many applications.

Convenient Packaging

  • Using up to 70% less plastic than a regular tip box, GilsonTM's reload pack is an eco-friendly and affordable solution.
  • Its small size makes it easy to store.
  • The racks are fully autoclavable and are color-coded for easy identification and compliance with Good Laboratory Practice (GLP).

Specifications

Volume (Metric) 0.1 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model DL10
Length (English) 1.77 in.
Length (Metric) 45 mm
Certifications/Compliance ISO 9001
For Use With (Application) Refill empty TIPACK™ boxes
Diameter (Metric) Outer .4 mm
Format Refill
Tip Style Wide Bore
Packaging Reload pack
Quantity 96 Tips/Pk.
Product Type Universal Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman Diamond
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less