Learn More
Gilson™ PIPETMANTM Comfort Handle
Designed to help reduce pipetting fatigue, the customizable Comfort Handle provides increased comfort to those currently using PIPETMAN Classic, NeoTM, and G pipettes
Supplier: Gilson™ F1614851
Description
- A large, more comfortable finger hook and ergonomically designed handle to fit better in your hand, PIPETMAN Comfort Handle offers a better grip to support your daily pipetting activity and aids in reducing the risk of Repetitive Strain Injuries (RSI).
- A dedicated identification zone has been integrated into the finger hook for quick and easy identification
- Autoclavable at 121°C (252°F), 1 atm for 20 minutes
- Includes removable, glossy, pre-cut adhesive labels that will work with any laser printer
Greener Choice Claims
- Environmental benefits include:
- Source Reduction
Specifications
Autoclavable | |
Glossy, pre-cut adhesive labels that will work with any laser printer | |
PIPETMAN Classic, Neo™, and G pipettes |
Help reduce pipetting fatigue of daily pipetting activity | |
Handle |