Learn More
Andwin Scientific Inc PIPETE LITE 8 CHAN 200UL DFS Item
Supplier:  Andwin Scientific Inc L8200XLS
The Multichannel Manual Pipet-Lite XLS, L8-200XLS+ is a manual 8-channel pipette that offers uniform sample pickup. This product has a light weight and together with low spring forces and tip ejection forces, users can utilize this pipette without any difficulty. The LTS tips and its new push-rod design give unparalleled accuracy and precision across all channels for certainly even sample pickup and dispensing. This product has a volume range of 20 L 200 L.
Catalog No. NC0403489
Only null left
Explore available promotions