missing translation for 'onlineSavingsMsg'
Learn More

Pipet Filler

Catalog No. S32244
Change view
Click to view available options
Color:
Black
Material:
Rubber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
S32244 Black Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S32244 Supplier Bal Supply LLC Supplier No. 5030BLACK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fits pipets up to 50mL.

  • Precise to 0.01mL
  • Chemical-resistant rubber
  • Expel air by pressing valve A on top of the bulb
  • Draw liquid up by pressing S valve on stem
  • To release, press valve E on side of the stem

Specifications

Autoclavable Not Autoclavable
Color Black
Material Rubber
Type Pipette Filler
For Use With (Equipment) Pipet