missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Pipet Cone

Catalog No. FB14955204
Change view
Click to view available options
Pipet Cone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
FB14955204 Pipet Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB14955204 Supplier Fisherbrand™ Supplier No. PH0111B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • For use with the Fisherbrand Pipet Controller
  • Autoclavable replacement cone

Specifications

Description Replacement parts only
Autoclavable Autoclavable
Color Blue
For Use With (Equipment) Fisherbrand Pipet Controller (FB14955202)