missing translation for 'onlineSavingsMsg'
Learn More

Piperonyloyl chloride, 98%

CAS: 25054-53-9 | C8H5ClO3 | 184.58 g/mol

$78.53 - $78.53

Chemical Identifiers

CAS 25054-53-9
Molecular Formula C8H5ClO3
Molecular Weight (g/mol) 184.58
MDL Number MFCD00016904
InChI Key ZRSGZIMDIHBXIN-UHFFFAOYSA-N
Synonym piperonyloyl chloride, benzo d 1,3 dioxole-5-carbonyl chloride, 2h-1,3-benzodioxole-5-carbonyl chloride, piperonoyl chloride, benzo 1,3 dioxole-5-carbonyl chloride, acmc-20ajvo, piperonylic acid chloride, 3,4-methylenedioxybenzoylchloride, 3,4-methylenedioxybenzoyl chloride
PubChem CID 2734749
IUPAC Name 1,3-benzodioxole-5-carbonyl chloride
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370790010
SDS View Documents
Thermo Scientific Chemicals
370790010
1 g Glass bottle
Each for $78.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25054-53-9
184.58
ZRSGZIMDIHBXIN-UHFFFAOYSA-N
2734749
C1OC2=C(O1)C=C(C=C2)C(=O)Cl
C8H5ClO3
MFCD00016904
piperonyloyl chloride, benzo d 1,3 dioxole-5-carbonyl chloride, 2h-1,3-benzodioxole-5-carbonyl chloride, piperonoyl chloride, benzo 1,3 dioxole-5-carbonyl chloride, acmc-20ajvo, piperonylic acid chloride, 3,4-methylenedioxybenzoylchloride, 3,4-methylenedioxybenzoyl chloride
1,3-benzodioxole-5-carbonyl chloride

Specifications

25054-53-9
Blue-Gray to White
97.5% min. (GC)
C8H5ClO3
1 g
ZRSGZIMDIHBXIN-UHFFFAOYSA-N
1,3-benzodioxole-5-carbonyl chloride
2734749
98%
Piperonyloyl chloride
77°C to 81°C
Authentic
Glass bottle
MFCD00016904
piperonyloyl chloride, benzo d 1,3 dioxole-5-carbonyl chloride, 2h-1,3-benzodioxole-5-carbonyl chloride, piperonoyl chloride, benzo 1,3 dioxole-5-carbonyl chloride, acmc-20ajvo, piperonylic acid chloride, 3,4-methylenedioxybenzoylchloride, 3,4-methylenedioxybenzoyl chloride
C1OC2=C(O1)C=C(C=C2)C(=O)Cl
184.58
184.58
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only