missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperonylonitrile 98.0+%, TCI America™
$86.64 - $169.41
Chemical Identifiers
| CAS | 4421-09-4 |
|---|---|
| Molecular Formula | C8H5NO2 |
| Molecular Weight (g/mol) | 147.13 |
| MDL Number | MFCD00005820 |
| InChI Key | PKRWWZCDLJSJIF-UHFFFAOYSA-N |
| Synonym | piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358 |
| PubChem CID | 78136 |
| IUPAC Name | 1,3-benzodioxole-5-carbonitrile |
| SMILES | C1OC2=C(O1)C=C(C=C2)C#N |
Chemical Identifiers
| 4421-09-4 | |
| 147.13 | |
| PKRWWZCDLJSJIF-UHFFFAOYSA-N | |
| 78136 | |
| C1OC2=C(O1)C=C(C=C2)C#N |
| C8H5NO2 | |
| MFCD00005820 | |
| piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358 | |
| 1,3-benzodioxole-5-carbonitrile |
Specifications
| 4421-09-4 | |
| C8H5NO2 | |
| 10 g | |
| PKRWWZCDLJSJIF-UHFFFAOYSA-N | |
| 1,3-benzodioxole-5-carbonitrile | |
| 78136 | |
| ≥98.0% (GC) | |
| Piperonylonitrile |
| 94°C | |
| MFCD00005820 | |
| piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358 | |
| C1OC2=C(O1)C=C(C=C2)C#N | |
| 147.13 | |
| 147.13 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TO2645000
TSCA : No