missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperonylonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 94°C |
| Quantity | 25 g |
| Formula Weight | 147.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Piperonylonitrile |
Safety and Handling
| RTECSNumber | TO2645000 |
| TSCA | No |