missing translation for 'onlineSavingsMsg'
Learn More

Piperonylic acid hydrazide, 97%

Catalog No. p-7033575
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5071303 1 g
AAH5071306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5071303 Supplier Thermo Scientific Chemicals Supplier No. H5071303
Only null left

CAS: 22026-39-7 | C8H8N2O3 | 180.163 g/mol

Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 22026-39-7
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00060505
InChI Key RAXBGBHBUFGWPG-UHFFFAOYSA-N
Synonym 3,4-methylenedioxybenzhydrazide, benzo d 1,3 dioxole-5-carbohydrazide, piperonylic acid hydrazide, 2h-1,3-benzodioxole-5-carbohydrazide, 3,4-methylenedioxybenzoic acid hydrazide, 3,4-methylenedioxy benzohydrazide, 1,3-benzodioxole-5-carboxylic acid, hydrazide, 3,4-methylenedioxy benzhydrazide, benzo 1,3 dioxole-5-carboxylic acid hydrazide, 2h-benzo d 1,3-dioxolane-5-carbohydrazide
PubChem CID 89158
IUPAC Name 1,3-benzodioxole-5-carbohydrazide
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)NN

Specifications

Melting Point 176°C to 178°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material Piperonylic acid hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only