Learn More
Piperonylic acid, 99%
CAS: 94-53-1 | C8H6O4 | 166.13 g/mol
$78.56 - $78.56
Chemical Identifiers
| CAS | 94-53-1 |
|---|---|
| Molecular Formula | C8H6O4 |
| Molecular Weight (g/mol) | 166.13 |
| MDL Number | MFCD00005830 |
| InChI Key | VDVJGIYXDVPQLP-UHFFFAOYSA-N |
| Synonym | piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid |
| PubChem CID | 7196 |
| IUPAC Name | 1,3-benzodioxole-5-carboxylic acid |
| SMILES | OC(=O)C1=CC=C2OCOC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC131370050
|
Thermo Scientific Chemicals
131370050 |
5 g | Glass bottle |
Each for $78.56
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 94-53-1 | |
| 166.13 | |
| VDVJGIYXDVPQLP-UHFFFAOYSA-N | |
| 7196 | |
| OC(=O)C1=CC=C2OCOC2=C1 |
| C8H6O4 | |
| MFCD00005830 | |
| piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid | |
| 1,3-benzodioxole-5-carboxylic acid |
Specifications
| 94-53-1 | |
| White to Beige | |
| 99% | |
| C8H6O4 | |
| 5 g | |
| 15, 7590 | |
| Solubility in water: slightly soluble. Other solubilities: slightly soluble in chloroform,cold alcohol and,ether | |
| OC(=O)C1=CC=C2OCOC2=C1 | |
| 166.13 | |
| 166.13 | |
| Powder |
| 229°C to 235°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005830 | |
| 19, 269 | |
| piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid | |
| VDVJGIYXDVPQLP-UHFFFAOYSA-N | |
| 1,3-benzodioxole-5-carboxylic acid | |
| 7196 | |
| 99% | |
| Piperonylic acid, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
EINECSNumber : 202-342-8
RTECSNumber : DF4912765
TSCA : TSCA
RUO – Research Use Only