Learn More
Piperonylic acid, 98+%
CAS: 94-53-1 | C8H6O4 | 166.13 g/mol
Supplier: Thermo Scientific Chemicals A1410522
Description
Piperonylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPiperonylic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Piperonylic acid | |
| 230°C to 235°C | |
| MFCD00005830 | |
| 150206 | |
| piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid | |
| VDVJGIYXDVPQLP-UHFFFAOYSA-N | |
| 1,3-benzodioxole-5-carboxylic acid | |
| 7196 | |
| ≥98% |
| 94-53-1 | |
| C8H6O4 | |
| 100 g | |
| 14,7477 | |
| Slightly soluble in water. | |
| OC(=O)C1=CC=C2OCOC2=C1 | |
| 166.13 | |
| 166.13 |
Chemical Identifiers
| 94-53-1 | |
| 166.13 | |
| VDVJGIYXDVPQLP-UHFFFAOYSA-N | |
| 7196 | |
| OC(=O)C1=CC=C2OCOC2=C1 |
| C8H6O4 | |
| MFCD00005830 | |
| piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid | |
| 1,3-benzodioxole-5-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
H302+H312+H332-H319
EINECSNumber : 202-342-8
RTECSNumber : DF4912765
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only