missing translation for 'onlineSavingsMsg'
Learn More

Piperonylic acid, 98+%

Catalog No. AAA1410536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1410536 500 g
AAA1410514 25 g
AAA1410522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1410536 Supplier Thermo Scientific Chemicals Supplier No. A1410536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94-53-1 | C8H6O4 | 166.13 g/mol

Piperonylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 94-53-1
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00005830
InChI Key VDVJGIYXDVPQLP-UHFFFAOYSA-N
Synonym piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid
PubChem CID 7196
IUPAC Name 1,3-benzodioxole-5-carboxylic acid
SMILES OC(=O)C1=CC=C2OCOC2=C1

Specifications

Melting Point 230°C to 235°C
Quantity 500 g
Beilstein 150206
Merck Index 14,7477
Solubility Information Slightly soluble in water.
Formula Weight 166.13
Percent Purity ≥98%
Chemical Name or Material Piperonylic acid
Hazard Category H302+H312+H332-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
EINECSNumber 202-342-8
RTECSNumber DF4912765
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only