missing translation for 'onlineSavingsMsg'
Learn More

Piperonylacetone 98.0+%, TCI America™

Catalog No. P108925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P108925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P108925G Supplier TCI America Supplier No. P108925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55418-52-5
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00016910
InChI Key TZJLGGWGVLADDN-UHFFFAOYSA-N
Synonym piperonyl acetone, piperonylacetone, 4-3,4-methylenedioxyphenyl-2-butanone, 4-1,3-benzodioxol-5-yl-2-butanone, 4-1,3-benzodioxol-5-yl butan-2-one, 2-butanone, 4-1,3-benzodioxol-5-yl, unii-081vvc9h34, fema no. 2701, 3,4-methylenedioxybenzylacetone, 4-benzo d 1,3 dioxol-5-yl butan-2-one
PubChem CID 62098
IUPAC Name 4-(1,3-benzodioxol-5-yl)butan-2-one
SMILES CC(=O)CCC1=CC2=C(C=C1)OCO2

Specifications

Melting Point 51°C
Color White
Boiling Point 165°C
Quantity 25 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Piperonylacetone