missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperonyl butoxide, tech. 90%
Description
Piperonyl butoxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperonyl butoxide is used as a pharmaceutical intermediate.
Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Piperonyl butoxide is used as a pharmaceutical intermediate.
Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.059 |
| Boiling Point | 155°C (0.3 mmHg) |
| Flash Point | 171°C (340°F) |
| Refractive Index | 1.498 |
| Quantity | 25 mL |
| UN Number | UN3082 |
| Beilstein | 288063 |
| Solubility Information | Soluble in benzene,ethanol and methanol. Not miscible or difficult to mix with water. |
| IUPAC Name | 5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole |
| Formula Weight | 338.44 |
| Show More |
Safety and Handling
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
| EINECSNumber | 200-076-7 |
| RTECSNumber | XS8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only