missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl butoxide, tech. 90%

Catalog No. AAH60771AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60771AC 25 mL
AAH60771AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60771AC Supplier Thermo Scientific Chemicals Supplier No. H60771AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51-03-6 | C19H30O5 | 338.44 g/mol

Piperonyl butoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonyl butoxide is used as a pharmaceutical intermediate.

Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 51-03-6
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
MDL Number MFCD00005842
InChI Key FIPWRIJSWJWJAI-UHFFFAOYSA-N
Synonym piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergist
PubChem CID 5794
ChEBI CHEBI:32687
SMILES CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC

Specifications

Density 1.059
Boiling Point 155°C (0.3 mmHg)
Flash Point 171°C (340°F)
Refractive Index 1.498
Quantity 25 mL
UN Number UN3082
Beilstein 288063
Solubility Information Soluble in benzene,ethanol and methanol. Not miscible or difficult to mix with water.
IUPAC Name 5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
Formula Weight 338.44
Percent Purity 90%
Grade Technical
Chemical Name or Material Piperonyl butoxide
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 200-076-7
RTECSNumber XS8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only