missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl Butoxide 90.0+%, TCI America™

Catalog No. P045825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P045825G 25 g
P0458500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P045825G Supplier TCI America Supplier No. P045825G

Chemical Identifiers

CAS 51-03-6
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
MDL Number MFCD00005842
InChI Key FIPWRIJSWJWJAI-UHFFFAOYSA-N
Synonym piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergist
PubChem CID 5794
ChEBI CHEBI:32687
IUPAC Name 5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
SMILES CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 g
UN Number 2810
Formula Weight 338.44
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Piperonyl Butoxide
RTECSNumber XS8050000
TSCA Yes