missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl butoxide, 90%, Tech.

Catalog No. p-185490
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC334161000 100 mL Glass bottle
AC334165000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC334161000 Supplier Thermo Scientific Chemicals Supplier No. 334161000
Only null left

CAS: 51-03-6 | C19H30O5 | 338.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-03-6
Molecular Formula C19H30O5
Molecular Weight (g/mol) 338.44
MDL Number MFCD00005842
InChI Key FIPWRIJSWJWJAI-UHFFFAOYSA-N
Synonym piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergist
PubChem CID 5794
ChEBI CHEBI:32687
IUPAC Name 5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole
SMILES CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC

Specifications

Color Yellow
Density 1.0600g/mL
Boiling Point 180°C (1.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4970 to 1.507
Quantity 100 mL
Beilstein 19, IV, 779
Specific Gravity 1.06
Merck Index 15, 7589
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with methanol,ethanol,benzene,freons,,and other organic solvents
Viscosity 40 mPa.s (25°C)
Formula Weight 338.44
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Piperonyl butoxide, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 200-076-7
RTECSNumber XS8050000
TSCA TSCA

RUO – Research Use Only